Perfluor-1-butansulfonsäurefluorid, Sigma-Aldrich®

Lieferant: Merck

Synonyme: Perfluoro-1-butanesulphonyl fluoride, Nonafluorobutanesulphonyl fluoride

M-Clarity™ Qualitätsstufe = MQ100

319732-100G 319732-25G
SIAL319732-100GEA 169 EUR
SIAL319732-100G SIAL319732-25G
Perfluor-1-butansulfonsäurefluorid, Sigma-Aldrich®
Perfluor-1-butansulfonsäurefluorid
Formel: C₄F₁₀O₂S
Molecular Weight: 302,09 g/mol
MDL: MFCD00007422
CAS-Nummer: 375-72-4
EINECS: 206-792-6
UN: 3265
ADR: 8,II

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR