(2-Fluor-3-pyridinyl)methanol

Lieferant: COMBI-BLOCKS

Synonyme: 2-Fluoro-3-(Hydroxymethyl)pyridine

SS-3006-1G SS-3006-5G
COBBSS-3006-1GEA 75.9 EUR
COBBSS-3006-1G COBBSS-3006-5G
(2-Fluor-3-pyridinyl)methanol
(2-Fluor-3-pyridinyl)methanol
Formel: C₆H₆FNO
Molecular Weight: 127,1200 g/mol
MDL: MFCD03092920
CAS-Nummer: 131747-55-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR