4-Chlor-1-iod-2-nitrobenzol

Lieferant: COMBI-BLOCKS

HC-2613-5G HC-2613-1G
COBBHC-2613-5GEA 207 EUR
COBBHC-2613-5G COBBHC-2613-1G
4-Chlor-1-iod-2-nitrobenzol
4-Chlor-1-iod-2-nitrobenzol
Formel: C₆H₃ClINO₂
Molecular Weight: 283,4500 g/mol
MDL: MFCD11226294
CAS-Nummer: 5446-05-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR