1-(2-Chlorethoxy)-4-nitrobenzol

Lieferant: BIONET RESEARCH

MS-5009.1 MS-5009.10 MS-5009.5 MS-5009.500
BIOTMS-5009.1EA 0 EUR
BIOTMS-5009.1 BIOTMS-5009.10 BIOTMS-5009.5 BIOTMS-5009.500
1-(2-Chlorethoxy)-4-nitrobenzol
1-(2-Chloroethoxy)-4-nitrobenzene
Formel: C₈H₈ClNO₃
Molecular Weight: 201.6100 g/mol
MDL: MFCD00674505
CAS-Nummer: 3383-72-0
EINECS: 425-790-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR