2,7-Dichlor-8-methylchinolin
Lieferant: COMBI-BLOCKS
PY-1308-1G
PY-1308-5G
COBBPY-1308-1GEA
204
EUR
COBBPY-1308-1G
COBBPY-1308-5G
2,7-Dichlor-8-methylchinolin
2,7-Dichloro-8-methylquinoline
Formel:
C₁₀H₇Cl₂N |
MDL:
MFCD15143625 CAS-Nummer: 1215205-97-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...