2-Chlor-8-methylchinolin

Lieferant: COMBI-BLOCKS

PY-1283-5G PY-1283-1G PY-1283-250M
COBBPY-1283-5GEA 614 EUR
COBBPY-1283-5G COBBPY-1283-1G COBBPY-1283-250M
2-Chlor-8-methylchinolin
2-Chloro-8-methylquinoline
Formel: C₁₀H₈ClN
Molecular Weight: 177,6300 g/mol
MDL: MFCD11108682
CAS-Nummer: 4225-85-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR