4,7-Dichlor-8-methylchinolin
Lieferant: COMBI-BLOCKS
HC-6484-5G
HC-6484-1G
COBBHC-6484-5GEA
451
EUR
COBBHC-6484-5G
COBBHC-6484-1G
4,7-Dichlor-8-methylchinolin
4,7-Dichloro-8-methylquinoline
Formel:
C₁₀H₇Cl₂N Molecular Weight: 212,800 g/mol |
MDL:
MFCD02684150 CAS-Nummer: 643039-79-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...