5-Chlor-8-methylchinolin

Lieferant: COMBI-BLOCKS

HC-2710-5G HC-2710-1G
COBBHC-2710-5GEA 873 EUR
COBBHC-2710-5G COBBHC-2710-1G
5-Chlor-8-methylchinolin
5-Chloro-8-methylquinoline
Formel: C₁₀H₈ClN
Molecular Weight: 177.63 g/mol
MDL: MFCD00463515
CAS-Nummer: 78941-95-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR