N,N-Dimethylcarbamoylchlorid, Sigma-Aldrich®
Lieferant: Merck
Synonyme:
Dimethylcarbamyl chloride
M-Clarity™ Qualitätsstufe = MQ200
D152803-100G
D152803-500G
D152803-5G
SIALD152803-100GEA
42.2
EUR
SIALD152803-100G
SIALD152803-500G
SIALD152803-5G
N,N-Dimethylcarbamoylchlorid, Sigma-Aldrich®
N,N-Dimethylcarbamoylchlorid
Formel:
C₃H₆ClNO |
MDL:
MFCD00000635 CAS-Nummer: 79-44-7 EINECS: 201-208-6 UN: 2262 ADR: 8,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...