5-Fluor-2'-deoxyuridin (Floxuridin), Sigma-Aldrich®
Lieferant: Merck
Synonyme: 5-Fluoro-1-((2R,4S,5R)-4-hydroxy-5-(hydroxymethyl)tetrahydrofuran-2-yl)pyrimidine-2,4(1H,3H)-dione, 5-Fluoro-2'-deoxyuridine, Floxuridine
F0503-100MG F0503-250MG F0503-1G
SIALF0503-100MGEA 289 EUR
SIALF0503-100MG SIALF0503-250MG SIALF0503-1G
5-Fluor-2'-deoxyuridin (Floxuridin), Sigma-Aldrich®
5-Fluor-2'-deoxyuridin (Floxuridin)
Formel: C₉H₁₁FN₂O₅
Molecular Weight: 246,19 g/mol
MDL: MFCD00006530
CAS-Nummer: 50-91-9
EINECS: 200-072-5
UN: 2811
ADR: 6.1,III