5-Brom-2'-deoxyuridin (Broxuridin), Sigma-Aldrich®
Lieferant: Merck
Synonyme: 5-Bromo-1-((2R,4S,5R)-4-hydroxy-5-(hydroxymethyl)tetrahydrofuran-2-yl)pyrimidine-2,4(1H,3H)-dione, 5-Bromo-2'-deoxyuridine, 5-BrdU
B9285-250MG B9285-50MG B9285-1G
SIALB9285-250MGEA 103 EUR
SIALB9285-250MG SIALB9285-50MG SIALB9285-1G
5-Brom-2'-deoxyuridin (Broxuridin), Sigma-Aldrich®
5-Brom-2'-deoxyuridin (Broxuridin)
Formel: C₉H₁₁BrN₂O₅
Molecular Weight: 307,10 g/mol
MDL: MFCD00006529
CAS-Nummer: 59-14-3
EINECS: 200-415-9