4-(Trifluormethyl)benzhydrol ≥97%

Lieferant: Thermo Scientific
Achtung

Synonyme: Phenyl(4-(trifluoromethyl)phenyl)methanol

L14147.03 L14147.06
L14147.03EA 16.1 EUR
L14147.03 L14147.06
4-(Trifluormethyl)benzhydrol ≥97%
4-(Trifluormethyl)benzhydrol

Purity: 97% CAS No.: 395-23-3 Molecular Formula: C14H11F3O Formula Weight: 252.23 Melting Point: 59-61° Boiling Point: 122-124°/0.4mm MDL No.: MFCD00014399

Formel: C₁₄H₁₁F₃O
Molecular Weight: 252,23 g/mol
Siedepunkt: 122…124 °C (0,4 mmHg)
Schmelzpunkt: 59…61 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00014399
CAS-Nummer: 395-23-3
EINECS: 209-917-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR