4-(Trifluormethyl)benzhydrol

Lieferant: FLUOROCHEM

Synonyme: Phenyl(4-(trifluoromethyl)phenyl)methanol

007592-5G 007592-1G
FLUO007592-5GEA 291 EUR
FLUO007592-5G FLUO007592-1G
4-(Trifluormethyl)benzhydrol
4-(Trifluormethyl)benzhydrol
Formel: C₁₄H₁₁F₃O
Molecular Weight: 252,23 g/mol
MDL: MFCD00014399
CAS-Nummer: 395-23-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR