Azidotrimethylsilan 94%
Lieferant: Thermo Scientific
|
Gefahr
|
Synonyme:
Trimethylsilyl azide
167930100
167930500
167932500
ACRO167930100EA
62.9
EUR
ACRO167930100
ACRO167930500
ACRO167932500
Azidotrimethylsilan 94%
Azidotrimethylsilane
Formel:
C₃H₉N₃Si Molecular Weight: 115,21 g/mol Siedepunkt: 95…96 °C (760 mmHg) Dichte: 0,876 g/cm³ Flammpunkt: 6 °C Lagertemperatur: Kühlschrank |
MDL:
MFCD00001986 CAS-Nummer: 4648-54-8 EINECS: 225-078-5 UN: 1992 ADR: 3,II |
Spezifikation Testergebnisse
Appearance | Clear to slightly hazy colorless Liquid |
Infrared spectrum | Conforms |
Refractive index | 1.4110 to 1.4180 (20°C, 589 nm) |
GC | ≥92 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...