Azidotrimethylsilan ≥94%

Lieferant: Thermo Scientific
Gefahr

Synonyme: Trimethylsilyl azide

L00173.06 L00173.14 L00173.22
L00173.06EA 30.4 EUR
L00173.06 L00173.14 L00173.22
Azidotrimethylsilan ≥94%
Azidotrimethylsilane

Purity: 94% CAS No.: 4648-54-8 Molecular Formula: C3H9N3Si Formula Weight: 115.21 Storage Temperature: 2°C to 8°C Flash Point: 6°(42°F) Refractive Index: 1.4140 Melting Point: -95° Boiling Point: 95-99° Beilstein Reference: 1903730 UN No.: 1992 MDL No.: MFCD00001986

Formel: C₃H₉N₃Si
Molecular Weight: 115,21 g/mol
Siedepunkt: 95…99 °C
Schmelzpunkt: –95 °C
Dichte: 0,872
Flammpunkt: 6 °C (42 °F)
MDL: MFCD00001986
CAS-Nummer: 4648-54-8
EINECS: 225-078-5
UN: 1992
ADR: 3,II

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR