1-(Trimethylsiloxy)-1,3-butadien

Lieferant: CHEMPUR SILANE

SILS18725.10 SILS18725.25
SILNSILS18725.10EA 0 EUR
SILNSILS18725.10 SILNSILS18725.25
1-(Trimethylsiloxy)-1,3-butadien
1-(Trimethylsiloxy)-1,3-butadien
Formel: C₇H₁₄OSi
CAS-Nummer: 6651-43-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR