2-(Trimethylsiloxy)-1,3-butadien

Lieferant: CHEMPUR SILANE

Synonyme: (Buta-1,3-dien-2-yloxy)trimethylsilane

SILS18750.5 SILS18750.25
SILNSILS18750.5EA 0 EUR
SILNSILS18750.5 SILNSILS18750.25
2-(Trimethylsiloxy)-1,3-butadien
2-(Trimethylsiloxy)-1,3-butadien
Formel: C₇H₁₄OSi
Molecular Weight: 142.27 g/mol
MDL: MFCD00009852
CAS-Nummer: 38053-91-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR