Diethyl(bromdifluormethyl)phosphonat

Lieferant: FLUOROCHEM

010248-5G 010248-25G
FLUO010248-5GEA 48.8 EUR
FLUO010248-5G FLUO010248-25G
Diethyl(bromdifluormethyl)phosphonat
Diethyl(bromdifluormethyl)phosphonat
Formel: C₅H₁₀BrF₂O₃P
Molecular Weight: 267.01 g/mol
MDL: MFCD00069118
CAS-Nummer: 65094-22-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR