N-(Ethoxycarbonyl)phthalimid, Sigma-Aldrich®

Lieferant: Merck

Synonyme: Ethyl 1,3-dioxoisoindoline 2-carboxylate, N-Carbethoxyphthalimide

M-Clarity™ Qualitätsstufe = MQ100

C5459-100G C5459-25G
SIALC5459-100GEA 112 EUR
SIALC5459-100G SIALC5459-25G
N-(Ethoxycarbonyl)phthalimid, Sigma-Aldrich®
N-(Ethoxycarbonyl)phthalimid
Formel: C₁₁H₉NO₄
Molecular Weight: 219,20 g/mol
MDL: MFCD00005893
CAS-Nummer: 22509-74-6
EINECS: 245-048-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR