6-Nitrocumarin ≥98%

Lieferant: Thermo Fisher Scientific

A14792.06 A14792.14
A14792.06EA 42.1 EUR
A14792.06 A14792.14
6-Nitrocumarin ≥98%
6-Nitrocumarin

Purity: 98+% CAS No.: 2725-81-7 Molecular Formula: C9H5NO4 Formula Weight: 191.14 Odour: Odourless Melting Point: 186-190° Beilstein Reference: 166079 MDL No.: MFCD00016973

Formel: C₉H₅NO₄
Schmelzpunkt: 186…190 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00016973
CAS-Nummer: 2725-81-7
EINECS: 220-341-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR