6-Nitrocumarin

Lieferant: Thermo Scientific

BTB12331.1 BTB12331EA BTB12331.50
MAYBBTB12331.1EA 0 EUR
MAYBBTB12331.1 MAYBBTB12331EA MAYBBTB12331.50
6-Nitrocumarin
6-Nitrocumarin
Formel: C₉H₅NO₄
MDL: MFCD00016973
CAS-Nummer: 2725-81-7
EINECS: 220-341-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR