7-Methoxy-2H-chromen-3-carbonsäureethylester

Lieferant: CHEMPUR

33-0049.1 33-0049.5
CHMP33-0049.1EA 0 EUR
CHMP33-0049.1 CHMP33-0049.5
7-Methoxy-2H-chromen-3-carbonsäureethylester
7-Methoxy-2H-chromen-3-carbonsäureethylester
Formel: C₁₃H₁₄O₄
MDL: MFCD04114608
CAS-Nummer: 885271-34-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR