6-Methoxy-2H-chromen-3-carbonsäureethylester

Lieferant: CHEMPUR

33-0043.1 33-0043.5
CHMP33-0043.1EA 0 EUR
CHMP33-0043.1 CHMP33-0043.5
6-Methoxy-2H-chromen-3-carbonsäureethylester
6-Methoxy-2H-chromen-3-carbonsäureethylester
Formel: C₁₃H₁₄O₄
MDL: MFCD01662448
CAS-Nummer: 57543-61-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR