8-Methoxy-2H-chromen-3-carbonsäure

Lieferant: BIONET RESEARCH

4G-319S.500 4G-319S.1 4G-319S.5 4G-319S.10
BIOT4G-319S.500EA 0 EUR
BIOT4G-319S.500 BIOT4G-319S.1 BIOT4G-319S.5 BIOT4G-319S.10
8-Methoxy-2H-chromen-3-carbonsäure
8-Methoxy-2H-chromen-3-carbonsäure
Formel: C₁₁H₁₀O₄
CAS-Nummer: 57543-59-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR