7-Methoxy-2H-chromen-3-carbonsäure

Lieferant: CHEMPUR

33-0046.1 33-0046.5
CHMP33-0046.1EA 0 EUR
CHMP33-0046.1 CHMP33-0046.5
7-Methoxy-2H-chromen-3-carbonsäure
7-Methoxy-2H-chromen-3-carbonsäure
Formel: C₁₁H₁₀O₄
MDL: MFCD04114606
CAS-Nummer: 57543-60-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR