6-Methoxy-2H-chromen-3-carbonsäure

Lieferant: BIONET RESEARCH

5G-302S.500 5G-302S.1 5G-302S.5 5G-302S.10
BIOT5G-302S.500EA 0 EUR
BIOT5G-302S.500 BIOT5G-302S.1 BIOT5G-302S.5 BIOT5G-302S.10
6-Methoxy-2H-chromen-3-carbonsäure
6-Methoxy-2H-chromen-3-carbonsäure
Formel: C₁₁H₁₀O₄
Molecular Weight: 206.1947 g/mol
MDL: MFCD00202057
CAS-Nummer: 57543-62-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR