7-(Diethylamino)-4-methylcumarin 99%

Lieferant: Thermo Scientific
Achtung

Synonyme: Coumarin 1, 7-(Diethylamino)-4-methyl-2H-chromen-2-one, 7-Diethylamino-4-methylcoumarin

B21938.22 B21938.36
B21938.22EA 90.5 EUR
B21938.22 B21938.36
7-(Diethylamino)-4-methylcumarin 99%
7-(Diethylamino)-4-methylcumarin

Purity: 99% CAS No.: 91-44-1 Molecular Formula: C14H17NO2 Formula Weight: 231.30 Melting Point: 72-75° Beilstein Reference: 193303

Formel: C₁₄H₁₇NO₂
Molecular Weight: 231,30 g/mol
Schmelzpunkt: 72…75 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00006864
CAS-Nummer: 91-44-1
EINECS: 202-068-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR