7-(Diethylamino)-4-methylcumarin, Sigma-Aldrich®

Lieferant: Merck

Synonyme: Coumarin 1, 7-(Diethylamino)-4-methyl-2H-chromen-2-one, 7-Diethylamino-4-methylcoumarin

D87759-100G D87759-500G D87759-5G
SIALD87759-100GEA 969 EUR
SIALD87759-100G SIALD87759-500G SIALD87759-5G
7-(Diethylamino)-4-methylcumarin, Sigma-Aldrich®
7-(Diethylamino)-4-methylcumarin
Formel: C₁₄H₁₇NO₂
Molecular Weight: 231,30 g/mol
MDL: MFCD00006864
CAS-Nummer: 91-44-1
EINECS: 202-068-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR