7-Amino-4-(methoxymethyl)cumarin

Lieferant: Thermo Scientific

RJF01106.1 RJF01106.10
MAYBRJF01106.1EA 0 EUR
MAYBRJF01106.1 MAYBRJF01106.10
7-Amino-4-(methoxymethyl)cumarin
7-Amino-4-(methoxymethyl)cumarin
Formel: C₁₁H₁₁NO₃
Molecular Weight: 205,22 g/mol
MDL: MFCD00068088
CAS-Nummer: 175205-10-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR