4-Methylumbelliferon ≥97%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
Synonyme:
7-Hydroxy-4-methylcumarin, Hymecromon
A10337.0B
A10337.18
A10337.30
A10337.0BEA
334
EUR
A10337.0B
A10337.18
A10337.30
4-Methylumbelliferon ≥97%
4-Methylumbelliferon
Purity: 97%
CAS No.: 90-33-5
Molecular Formula: C10H8O3
Formula Weight: 176.17
Melting Point: 186-191°
Solubility: Soluble in alcohol. Insoluble in cold water
Beilstein Reference: 142217
Merck Reference: 14,4854
MDL No.: MFCD00006866
Formel:
C₁₀H₈O₃ Molecular Weight: 176.17 g/mol Schmelzpunkt: 186…191 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00006866 CAS-Nummer: 90-33-5 EINECS: 201-986-7 |