7-Amino-4-(trifluormethyl)cumarin 99%
Lieferant: Thermo Scientific
|
Achtung
|
Synonyme:
7-Amino-4-(trifluoromethyl)coumarin, 7-Amino-4-trifluoromethylcoumarin, 7-Amino-4-(trifluoromethyl)-2H-chromen-2-one, Coumarin 151
A18400.01
A18400.03
A18400.01EA
38
EUR
A18400.01
A18400.03
7-Amino-4-(trifluormethyl)cumarin 99%
7-Amino-4-(trifluormethyl)cumarin
Purity: 99%
CAS No.: 53518-15-3
Molecular Formula: C10H6F3NO2
Formula Weight: 229.16
Melting Point: 222-223°
Beilstein Reference: 4456797
Formel:
C₁₀H₆F₃NO₂ Molecular Weight: 229,16 g/mol Schmelzpunkt: 222…223 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00006858 CAS-Nummer: 53518-15-3 EINECS: 258-599-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...