7-Amino-4-(trifluormethyl)cumarin

Lieferant: Thermo Scientific

Synonyme: 7-Amino-4-(trifluoromethyl)coumarin, 7-Amino-4-trifluoromethylcoumarin, 7-Amino-4-(trifluoromethyl)-2H-chromen-2-one, Coumarin 151

RJF00052.1 RJF00052.10
MAYBRJF00052.1EA 0 EUR
MAYBRJF00052.1 MAYBRJF00052.10
7-Amino-4-(trifluormethyl)cumarin
7-Amino-4-(trifluormethyl)cumarin
Formel: C₁₀H₆F₃NO₂
Molecular Weight: 229,16 g/mol
MDL: MFCD00006858
CAS-Nummer: 53518-15-3
EINECS: 258-599-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR