2-(Diethylamino)ethanthiol Hydrochlorid, Sigma-Aldrich®
Lieferant: Merck
M-Clarity™ Qualitätsstufe = MQ100
D86604-100G
D86604-25G
SIALD86604-100GEA
234
EUR
SIALD86604-100G
SIALD86604-25G
2-(Diethylamino)ethanthiol Hydrochlorid, Sigma-Aldrich®
2-(Diethylamino)ethanthiol Hydrochlorid
Formel:
C₆H₁₆ClNS |
CAS-Nummer:
1942-52-5 EINECS: 217-731-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...