2-(Dimethylamino)ethanthiol Hydrochlorid, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ200

D141003-100G D141003-25G
SIALD141003-100GEA 603 EUR
SIALD141003-100G SIALD141003-25G
2-(Dimethylamino)ethanthiol Hydrochlorid, Sigma-Aldrich®
2-(Dimethylamino)ethanthiol Hydrochlorid
Formel: C₄H₁₂ClNS
MDL: MFCD00012633
CAS-Nummer: 13242-44-9
EINECS: 236-221-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR