Imidazol-4,5-dicarbonsäure
Lieferant: FLUOROCHEM
Synonyme:
1H-Imidazole-4,5-dicarboxylic acid, 4,5-Imidazoledicarboxylic acid
009677-10G
009677-25G
FLUO009677-10GEA
30.5
EUR
FLUO009677-10G
FLUO009677-25G
Imidazol-4,5-dicarbonsäure
Imidazol-4,5-dicarbonsäure
Formel:
C₅H₄N₂O₄ Molecular Weight: 156,10 g/mol |
MDL:
MFCD00005200 CAS-Nummer: 570-22-9 EINECS: 209-327-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...