1-(p-Toluolsulfonyl)imidazol ≥98%
Lieferant: Thermo Scientific
|
Achtung
|
Synonyme:
1-(P-Toluenesulphonyl)imidazole, 1-Tosylimidazole
H54191.06
H54191.14
H54191.22
H54191.06EA
58.4
EUR
H54191.06
H54191.14
H54191.22
1-(p-Toluolsulfonyl)imidazol ≥98%
1-(p-Toluolsulfonyl)imidazol
Purity: 98+%
CAS No.: 2232-08-8
Molecular Formula: C10H10N2O2S
Formula Weight: 222.26
Melting Point: 76-78°
Beilstein Reference: 612451
Formel:
C₁₀H₁₀N₂O₂S Molecular Weight: 222,26 g/mol Schmelzpunkt: 76…78 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00005285 CAS-Nummer: 2232-08-8 EINECS: 218-771-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...