1-Methylimidazol-5-carbonsäuremethylester

Lieferant: COMBI-BLOCKS

Synonyme: Methyl 1-methylimidazole-5-carboxylate, Methyl 1-methyl-1H-imidazole-5-carboxylate

HI-1103-5G HI-1103-25G HI-1103-1G
COBBHI-1103-5GEA 188 EUR
COBBHI-1103-5G COBBHI-1103-25G COBBHI-1103-1G
1-Methylimidazol-5-carbonsäuremethylester
1-Methylimidazol-5-carbonsäuremethylester
Formel: C₆H₈N₂O₂
Molecular Weight: 140,14 g/mol
MDL: MFCD01567300
CAS-Nummer: 17289-20-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR