1-Methylimidazol-5-carbonsäuremethylester

Lieferant: Thermo Scientific

Synonyme: Methyl 1-methylimidazole-5-carboxylate, Methyl 1-methyl-1H-imidazole-5-carboxylate

TL00659DA TL00659EA
MAYBTL00659DAEA 0 EUR
MAYBTL00659DA MAYBTL00659EA
1-Methylimidazol-5-carbonsäuremethylester
1-Methylimidazol-5-carbonsäuremethylester
Formel: C₆H₈N₂O₂
Molecular Weight: 140,14 g/mol
MDL: MFCD01567300
CAS-Nummer: 17289-20-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR