2,3-Dihydrobenzo[b]furan-5-carbonsäure

Lieferant: FLUOROCHEM

Synonyme: 2,3-Dihydrobenzo[b]furan-5-carboxylicacid

CFF501.1 CFF501.5 017509-1G 017509-5G 017509-25G
CHMPCFF501.1EA 268 EUR
CHMPCFF501.1 CHMPCFF501.5 FLUO017509-1G FLUO017509-5G FLUO017509-25G
2,3-Dihydrobenzo[b]furan-5-carbonsäure
2,3-Dihydrobenzo[b]furan-5-carbonsäure
Formel: C₉H₈O₃
Molecular Weight: 164.1580 g/mol
MDL: MFCD00085012
CAS-Nummer: 76429-73-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR