2,3-Dihydrobenzo[b]furan-5-carbonsäure
Lieferant: Thermo Scientific
Synonyme:
2,3-Dihydrobenzo[b]furan-5-carboxylicacid
CC00801.250
CC00801.1
MAYBCC00801.250EA
0
EUR
MAYBCC00801.250
MAYBCC00801.1
2,3-Dihydrobenzo[b]furan-5-carbonsäure
2,3-Dihydrobenzo[b]furan-5-carbonsäure
Formel:
C₉H₈O₃ Molecular Weight: 164.1580 g/mol |
MDL:
MFCD00085012 CAS-Nummer: 76429-73-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...