2,3-Dihydrobenzo[b]furan-7-carbonsäure
Lieferant: FLUOROCHEM
Synonyme:
2,3-Dihydrobenzofuran-7-carboxylic acid
011337-1G
011337-5G
FLUO011337-1GEA
213
EUR
FLUO011337-1G
FLUO011337-5G
2,3-Dihydrobenzo[b]furan-7-carbonsäure
2,3-Dihydrobenzo[b]furan-7-carbonsäure
Formel:
C₉H₈O₃ Molecular Weight: 164,1580 g/mol |
MDL:
MFCD00671526 CAS-Nummer: 35700-40-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...