2-Butyl-5-chlor-6-nitrobenzoxazol

Lieferant: BIONET RESEARCH

7X-5092.500 7X-5092.1 7X-5092.5 7X-5092.10
BIOT7X-5092.500EA 0 EUR
BIOT7X-5092.500 BIOT7X-5092.1 BIOT7X-5092.5 BIOT7X-5092.10
2-Butyl-5-chlor-6-nitrobenzoxazol
2-Butyl-5-chloro-6-nitrobenzoxazole
Formel: C₁₁H₁₂N₂O₃
CAS-Nummer: 886360-94-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR