2-Butyl-1,3-benzoxazol-5-amin

Lieferant: BIONET RESEARCH

Synonyme: 5-Amino-2-(but-1-yl)-1,3-benzoxazole

7Y-0705.1 7Y-0705.10 7Y-0705.5 7Y-0705.500
BIOT7Y-0705.1EA 0 EUR
BIOT7Y-0705.1 BIOT7Y-0705.10 BIOT7Y-0705.5 BIOT7Y-0705.500
2-Butyl-1,3-benzoxazol-5-amin
2-Butyl-1,3-benzoxazol-5-amin
Formel: C₁₁H₁₄N₂O
CAS-Nummer: 885949-91-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR