2-Ethyl-1,3-benzoxazol-5-amin

Lieferant: BIONET RESEARCH

Synonyme: 5-Amino-2-ethyl-1,3-benzoxazole

2W-0838.500 2W-0838.1 2W-0838.5 2W-0838.10
BIOT2W-0838.500EA 0 EUR
BIOT2W-0838.500 BIOT2W-0838.1 BIOT2W-0838.5 BIOT2W-0838.10
2-Ethyl-1,3-benzoxazol-5-amin
2-Ethyl-1,3-benzoxazol-5-amin
Formel: C₉H₁₀N₂O
CAS-Nummer: 204771-75-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR