2-Methyl-1,3-benzoxazol-5-amin

Lieferant: BIONET RESEARCH

5X-0708.500 5X-0708.1 5X-0708.5 5X-0708.10
BIOT5X-0708.500EA 0 EUR
BIOT5X-0708.500 BIOT5X-0708.1 BIOT5X-0708.5 BIOT5X-0708.10
2-Methyl-1,3-benzoxazol-5-amin
2-Methyl-1,3-benzoxazol-5-amin
Formel: C₈H₈N₂O
MDL: MFCD00628970
CAS-Nummer: 72745-76-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR