2-Butyl-1,3-benzoxazol-6-amin

Lieferant: BIONET RESEARCH

Synonyme: 6-Amino-2-(but-1-yl)-1,3-benzoxazole

7Y-0706.5 7Y-0706.10 7Y-0706.500 7Y-0706.1
BIOT7Y-0706.5EA 0 EUR
BIOT7Y-0706.5 BIOT7Y-0706.10 BIOT7Y-0706.500 BIOT7Y-0706.1
2-Butyl-1,3-benzoxazol-6-amin
2-Butyl-1,3-benzoxazol-6-amin
Formel: C₁₁H₁₄N₂O
CAS-Nummer: 875850-00-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR