2-Ethyl-1,3-benzoxazol-6-amin

Lieferant: BIONET RESEARCH

1W-0924.500 1W-0924.1 1W-0924.5 1W-0924.10
BIOT1W-0924.500EA 0 EUR
BIOT1W-0924.500 BIOT1W-0924.1 BIOT1W-0924.5 BIOT1W-0924.10
2-Ethyl-1,3-benzoxazol-6-amin
2-Ethyl-1,3-benzoxazol-6-amin
Formel: C₉H₁₀N₂O
CAS-Nummer: 855423-35-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR