2-Methyl-1,3-benzoxazol-6-amin

Lieferant: BIONET RESEARCH

Synonyme: 2-Methylbenzo[d]oxazol-6-amine

1W-0848.500 1W-0848.1 1W-0848.5 1W-0848.10
BIOT1W-0848.500EA 0 EUR
BIOT1W-0848.500 BIOT1W-0848.1 BIOT1W-0848.5 BIOT1W-0848.10
2-Methyl-1,3-benzoxazol-6-amin
2-Methyl-1,3-benzoxazol-6-amin
Formel: C₁₁H₁₄N₂O
Molecular Weight: 148,1619 g/mol
MDL: MFCD03791181
CAS-Nummer: 5676-60-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR