2-Mercaptobenzoxazol ≥98%
Lieferant: Thermo Scientific
|
Achtung
|
Synonyme:
Benzo[d]oxazole-2-thiol
A12346.14
A12346.22
A12346.14EA
35.2
EUR
A12346.14
A12346.22
2-Mercaptobenzoxazol ≥98%
2-Mercaptobenzoxazole
Purity: 98+%
CAS No.: 2382-96-9
Molecular Formula: C7H5NOS
Formula Weight: 151.19
Melting Point: 189-194°
Beilstein Reference: 119483
Formel:
C₇H₅NOS Molecular Weight: 151,19 g/mol Schmelzpunkt: 189…194 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00005769 CAS-Nummer: 2382-96-9 EINECS: 219-191-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...