2-Mercaptobenzoxazol

Lieferant: CHEMPUR

Synonyme: Benzo[d]oxazole-2-thiol

SB01024.1 SB01024.10 SB01024.50
CHMPSB01024.1EA 0 EUR
CHMPSB01024.1 CHMPSB01024.10 CHMPSB01024.50
2-Mercaptobenzoxazol
2-Mercaptobenzoxazole
Formel: C₇H₅NOS
Molecular Weight: 151,19 g/mol
MDL: MFCD00005769
CAS-Nummer: 2382-96-9
EINECS: 219-191-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR